2-Fluor-5-nitrophenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
PC3014-25G
PC3014-5G
PC3014-100G
APOSPC3014-25GEA
0
EUR
APOSPC3014-25G
APOSPC3014-5G
APOSPC3014-100G
2-Fluor-5-nitrophenylboronsäure
2-Fluor-5-nitrophenylboronsäure
Formel:
C₆H₅BFNO₄ Lagertemperatur: Kühlschrank |
MDL:
MFCD03092929 CAS-Nummer: 819849-20-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...