2-(2,3,4-Trifluorphenyl)ethanol 98%

Lieferant: Apollo Scientific

PC302000-1G PC302000-5G
APOSPC302000-1GEA 125 EUR
APOSPC302000-1G APOSPC302000-5G
2-(2,3,4-Trifluorphenyl)ethanol 98%
2-(2,3,4-Trifluorphenyl)ethanol
Formel: C₈H₇F₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD04972479
CAS-Nummer: 886761-76-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR