2-(2,4,5-Trifluorphenyl)ethanol 97%

Lieferant: Apollo Scientific

PC302001-1G PC302001-5G PC302001-25G
APOSPC302001-1GEA 30.6 EUR
APOSPC302001-1G APOSPC302001-5G APOSPC302001-25G
2-(2,4,5-Trifluorphenyl)ethanol 97%
2-(2,4,5-Trifluorphenyl)ethanol
Formel: C₈H₇F₃O
Molecular Weight: 176,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660356
CAS-Nummer: 883267-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR