2-(2,5-Difluorphenyl)ethanol 97%

Lieferant: Apollo Scientific

PC302002-1G PC302002-5G PC302002-25G
APOSPC302002-1GEA 72.5 EUR
APOSPC302002-1G APOSPC302002-5G APOSPC302002-25G
2-(2,5-Difluorphenyl)ethanol 97%
2-(2,5-Difluorphenyl)ethanol
Formel: C₈H₈F₂O
Molecular Weight: 158.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00142840
CAS-Nummer: 862255-56-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR