(2,3-Dichlor-6-fluorphenyl)essigsäure 97%
Lieferant: Apollo Scientific
PC302007-1G
PC302007-5G
APOSPC302007-1GEA
105
EUR
APOSPC302007-1G
APOSPC302007-5G
(2,3-Dichlor-6-fluorphenyl)essigsäure 97%
(2,3-Dichlor-6-fluorphenyl)essigsäure
Formel:
(C₆H₂Cl₂F)CH₂COOH Molecular Weight: 223.03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115875 CAS-Nummer: 886497-57-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...