2,3-Difluor-4-methoxybenzylamin 95%

Lieferant: Apollo Scientific
Gefahr

PC302008-1G PC302008-5G
APOSPC302008-1GEA 143 EUR
APOSPC302008-1G APOSPC302008-5G
2,3-Difluor-4-methoxybenzylamin 95%
2,3-Difluor-4-methoxybenzylamin
Formel: C₈H₉F₂NO
Molecular Weight: 173,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115987
CAS-Nummer: 886500-75-0
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR