2-[2-Fluoro-4-(trifluoromethyl)phenyl]ethanol 98%
Lieferant: Apollo Scientific
PC302018-1G
PC302018-5G
APOSPC302018-1GEA
144
EUR
APOSPC302018-1G
APOSPC302018-5G
2-[2-Fluoro-4-(trifluoromethyl)phenyl]ethanol 98%
2-[2-Fluoro-4-(trifluoromethyl)phenyl]ethanol
Formel:
C₉H₈F₄O Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660357 CAS-Nummer: 886761-78-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...