3,5-Difluor-4-methoxyphenol 98%
Lieferant: Apollo Scientific
Synonyme:
1,3-Difluor-5-hydroxy-2-methoxybenzol, 2,6-Difluor-4-hydroxyanisol
PC302037-1G
PC302037-5G
PC302037-25G
APOSPC302037-1GEA
94.4
EUR
APOSPC302037-1G
APOSPC302037-5G
APOSPC302037-25G
3,5-Difluor-4-methoxyphenol 98%
3,5-Difluor-4-methoxyphenol
Formel:
C₇H₆F₂O₂ Schmelzpunkt: 67…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115918 CAS-Nummer: 443-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...