3-Chlor-5-(trifluormethoxy)phenol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Chlor-3-hydroxy-5-(trifluormethoxy)benzol
PC302050-1G
PC302050-5G
PC302050-10G
APOSPC302050-1GEA
102
EUR
APOSPC302050-1G
APOSPC302050-5G
APOSPC302050-10G
3-Chlor-5-(trifluormethoxy)phenol 98%
3-Chlor-5-(trifluormethoxy)phenol
Formel:
C₇H₄ClF₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832330 CAS-Nummer: 1017778-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...