4-Chlor-3-(trifluormethoxy)phenol 98%
Lieferant: Apollo Scientific
Synonyme:
1-Chlor-4-hydroxy-2-(trifluormethoxy)benzol
PC302060-1G
PC302060-5G
PC302060-25G
APOSPC302060-1GEA
60.9
EUR
APOSPC302060-1G
APOSPC302060-5G
APOSPC302060-25G
4-Chlor-3-(trifluormethoxy)phenol 98%
4-Chlor-3-(trifluormethoxy)phenol
Formel:
C₇H₄ClF₃O₂ Molecular Weight: 212.56 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660247 CAS-Nummer: 886500-85-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...