2-Fluor-p-anisidin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Fluor-4-anisidin, 4-Amino-3-fluoranisol, 2-Fluor-4-methoxyanilin
PC302074-1G
PC302074-5G
PC302074-10G
APOSPC302074-1GEA
21.9
EUR
APOSPC302074-1G
APOSPC302074-5G
APOSPC302074-10G
2-Fluor-p-anisidin 98%
2-Fluor-p-anisidin
Formel:
C₇H₈FNO Molecular Weight: 141,15 g/mol Schmelzpunkt: 48…51 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00047827 CAS-Nummer: 458-52-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...