2-Fluor-5-methylbenzylamin 98%

Lieferant: Apollo Scientific
Gefahr

PC302076-1G PC302076-5G PC302076-10G
APOSPC302076-1GEA 116 EUR
APOSPC302076-1G APOSPC302076-5G APOSPC302076-10G
2-Fluor-5-methylbenzylamin 98%
2-Fluor-5-methylbenzylamin
Formel: C₈H₁₀FN
Molecular Weight: 139.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06213371
CAS-Nummer: 93071-82-0
UN: 2735
ADR: 8 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR