2-Chlor-3-(trifluormethyl)phenylacetonitril 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-4-(trifluoromethyl)phenylacetonitrile

PC302090-1G PC302090-5G
APOSPC302090-1GEA 219 EUR
APOSPC302090-1G APOSPC302090-5G
2-Chlor-3-(trifluormethyl)phenylacetonitril 97%
2-Chlor-3-(trifluormethyl)phenylacetonitril
Formel: C₉H₅ClF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD09924313
CAS-Nummer: 1000530-58-4
UN: 3276
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR