3-Fluor-2-methylbenzylamin 98%

Lieferant: Apollo Scientific

PC302092-1G PC302092-5G PC302092-10G
APOSPC302092-1GEA 68.3 EUR
APOSPC302092-1G APOSPC302092-5G APOSPC302092-10G
3-Fluor-2-methylbenzylamin 98%
3-Fluor-2-methylbenzylamin
Formel: C₈H₁₀FN
Molecular Weight: 139,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06213676
CAS-Nummer: 771573-36-5
UN: 1760
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR