3-Fluor-5-methylbenzylamin 98%

Lieferant: Apollo Scientific

PC302093-1G PC302093-5G PC302093-10G
APOSPC302093-1GEA 84.1 EUR
APOSPC302093-1G APOSPC302093-5G APOSPC302093-10G
3-Fluor-5-methylbenzylamin 98%
3-Fluor-5-methylbenzylamin
Formel: C₈H₁₀FN
Molecular Weight: 139,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06213636
CAS-Nummer: 771573-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR