3-Methoxy-2-(trifluormethyl)benzoesäure 98%

Lieferant: Apollo Scientific
Achtung

PC302095-1G PC302095-5G
APOSPC302095-1GEA 101 EUR
APOSPC302095-1G APOSPC302095-5G
3-Methoxy-2-(trifluormethyl)benzoesäure 98%
3-Methoxy-2-(trifluoromethyl)benzoic acid
Formel: C₉H₇F₃O₃
Molecular Weight: 220,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185681
CAS-Nummer: 1214384-93-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR