2,4,6-Trifluor-3-methoxybenzoesäure 97%

Lieferant: Apollo Scientific

Synonyme: 3-Methoxy-2,4,6-trifluorobenzoic acid

PC302097-1G PC302097-5G
APOSPC302097-1GEA 69.6 EUR
APOSPC302097-1G APOSPC302097-5G
2,4,6-Trifluor-3-methoxybenzoesäure 97%
2,4,6-Trifluor-3-methoxybenzoesäure
Formel: C₉H₇F₃O₃
Molecular Weight: 220.15 g/mol
Schmelzpunkt: 100…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06660225
CAS-Nummer: 886499-94-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR