4-Chlor-3-fluorbenzamid

Lieferant: Apollo Scientific

PC302107-25G PC302107-5G PC302107-10G
APOSPC302107-25GEA 435 EUR
APOSPC302107-25G APOSPC302107-5G APOSPC302107-10G
4-Chlor-3-fluorbenzamid
4-Chlor-3-fluorbenzamid
Formel: C₇H₅ClFNO
Lagertemperatur: Raumtemperatur
CAS-Nummer: 874781-07-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR