4-Ethoxy-2,6-difluorobenzoyl chloride 97%

Lieferant: Apollo Scientific

Synonyme: 4-Ethoxy-2,6-difluorobenzoyl chloride

PC302109-1G PC302109-5G
APOSPC302109-1GEA 87 EUR
APOSPC302109-1G APOSPC302109-5G
4-Ethoxy-2,6-difluorobenzoyl chloride 97%
4-Ethoxy-2,6-difluorobenzoyl chloride
Formel: C₉H₇ClF₂O₂
Molecular Weight: 220,60 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22201053
CAS-Nummer: 1373920-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR