(4-Fluor-2-methylphenyl)essigsäure 98%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Fluoro-2-methylphenyl)acetic acid

PC302112-1G PC302112-5G PC302112-25G
APOSPC302112-1GEA 29 EUR
APOSPC302112-1G APOSPC302112-5G APOSPC302112-25G
(4-Fluor-2-methylphenyl)essigsäure 98%
(4-Fluor-2-methylphenyl)essigsäure
Formel: C₉H₉FO₂
Molecular Weight: 168.17 g/mol
Schmelzpunkt: 96…99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00049318
CAS-Nummer: 407640-40-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR