4-Fluoro-3-methyl-5-(trifluoromethyl)phenol 98%
Lieferant: Apollo Scientific
Synonyme:
4-Fluoro-3-methyl-5-(trifluoromethyl)phenol
PC302114-1G
PC302114-5G
APOSPC302114-1GEA
97.3
EUR
APOSPC302114-1G
APOSPC302114-5G
4-Fluoro-3-methyl-5-(trifluoromethyl)phenol 98%
4-Fluoro-3-methyl-5-(trifluoromethyl)phenol
Formel:
C₈H₆F₄O Molecular Weight: 194,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22201069 CAS-Nummer: 1373920-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...