(4-Fluor-3-methylphenyl)essigsäure 98%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Fluoro-3-methylphenyl)acetic acid

PC302116-1G PC302116-5G PC302116-10G
APOSPC302116-1GEA 55.1 EUR
APOSPC302116-1G APOSPC302116-5G APOSPC302116-10G
(4-Fluor-3-methylphenyl)essigsäure 98%
(4-Fluor-3-methylphenyl)essigsäure
Formel: C₉H₉FO₂
Molecular Weight: 168,17 g/mol
Schmelzpunkt: 105…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09832241
CAS-Nummer: 1000520-92-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR