5'-Fluor-2'-methylacetophenon 98%

Lieferant: Apollo Scientific

Synonyme: 5-Fluoro-2-methylacetophenone, 1-(5-Fluoro-2-methylphenyl)ethan-1-one

PC302118-25G PC302118-5G PC302118-1G
APOSPC302118-25GEA 359 EUR
APOSPC302118-25G APOSPC302118-5G APOSPC302118-1G
5'-Fluor-2'-methylacetophenon 98%
5'-Fluor-2'-methylacetophenon
Formel: C₉H₉FO
Molecular Weight: 152.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660189
CAS-Nummer: 29427-49-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR