2,4-Dibrom-5-(trifluormethoxy)anisol 97%

Lieferant: Apollo Scientific

PC302156-25G PC302156-5G PC302156-1G
APOSPC302156-25GEA 360 EUR
APOSPC302156-25G APOSPC302156-5G APOSPC302156-1G
2,4-Dibrom-5-(trifluormethoxy)anisol 97%
2,4-Dibrom-5-(trifluormethoxy)anisol
Formel: C₈H₅Br₂F₃O₂
Molecular Weight: 349.93 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22201036
CAS-Nummer: 1373920-68-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR