2',5'-Bis(trifluormethyl)acetophenon 97%
Lieferant: Apollo Scientific
Synonyme:
1-[2,5-Bis(trifluormethyl)phenyl]ethan-1-on
PC302159-25G
PC302159-5G
PC302159-1G
APOSPC302159-25GEA
316
EUR
APOSPC302159-25G
APOSPC302159-5G
APOSPC302159-1G
2',5'-Bis(trifluormethyl)acetophenon 97%
2',5'-Bis(trifluormethyl)acetophenon
Formel:
C₁₀H₆F₆O Molecular Weight: 256,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04116044 CAS-Nummer: 545410-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...