α-Brom-2-chlor-4,5-difluortoluol 97%

Lieferant: Apollo Scientific
Gefahr

PC302167-25G PC302167-5G PC302167-1G
APOSPC302167-25GEA 496 EUR
APOSPC302167-25G APOSPC302167-5G APOSPC302167-1G
α-Brom-2-chlor-4,5-difluortoluol 97%
α-Brom-2-chlor-4,5-difluortoluol
Formel: C₇H₄BrClF₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06660158
CAS-Nummer: 874285-21-9
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR