(2-Chlor-6-(trifluormethyl)phenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

PC302168-1G PC302168-5G
APOSPC302168-1GEA 105 EUR
APOSPC302168-1G APOSPC302168-5G
(2-Chlor-6-(trifluormethyl)phenyl)essigsäure 98%
(2-Chlor-6-(trifluormethyl)phenyl)essigsäure
Formel: C₉H₆ClF₃O₂
Molecular Weight: 238.59 g/mol
Schmelzpunkt: 149…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06660234
CAS-Nummer: 886500-31-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR