2-Methyl-5-(trifluoromethoxy)phenol 98%
Lieferant: Apollo Scientific
Synonyme:
2-methyl-5-(trifluoromethoxy)phenol
PC302180-1G
PC302180-5G
APOSPC302180-1GEA
126
EUR
APOSPC302180-1G
APOSPC302180-5G
2-Methyl-5-(trifluoromethoxy)phenol 98%
2-Methyl-5-(trifluoromethoxy)phenol
Formel:
C₉H₇F₃O₂ Molecular Weight: 192,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18399811 CAS-Nummer: 1261747-88-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...