3-Chlor-2-fluor-5-(trifluormethyl)anilin 97%

Lieferant: Apollo Scientific
Gefahr

PC302192-1G PC302192-5G PC302192-10G
APOSPC302192-1GEA 126 EUR
APOSPC302192-1G APOSPC302192-5G APOSPC302192-10G
3-Chlor-2-fluor-5-(trifluormethyl)anilin 97%
3-Chlor-2-fluor-5-(trifluormethyl)anilin
Formel: C₇H₄ClF₄N
Molecular Weight: 213.56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11519322
CAS-Nummer: 126538-85-0
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR