(3-Chlor-4-(trifluormethyl)phenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

PC302195-1G PC302195-5G PC302195-10G
APOSPC302195-1GEA 121 EUR
APOSPC302195-1G APOSPC302195-5G APOSPC302195-10G
(3-Chlor-4-(trifluormethyl)phenyl)essigsäure 98%
(3-Chlor-4-(trifluormethyl)phenyl)essigsäure
Formel: C₉H₆ClF₃O₂
Molecular Weight: 238,59 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1000568-54-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR