(3-Chlor-5-(trifluormethyl)phenyl)essigsäure 98%
Lieferant: Apollo Scientific
Synonyme:
2-(3-Chloro-5-(trifluoromethyl)phenyl)acetic acid
PC302198-1G
PC302198-5G
PC302198-10G
APOSPC302198-1GEA
37.7
EUR
APOSPC302198-1G
APOSPC302198-5G
APOSPC302198-10G
(3-Chlor-5-(trifluormethyl)phenyl)essigsäure 98%
(3-Chlor-5-(trifluormethyl)phenyl)essigsäure
Formel:
C₉H₆ClF₃O₂ Molecular Weight: 238.59 g/mol Schmelzpunkt: 125…127 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115851 CAS-Nummer: 886496-99-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...