2'-Chlor-3'-(trifluormethyl)acetophenon 97%

Lieferant: Apollo Scientific

PC302226-5G PC302226-1G
APOSPC302226-5GEA 241 EUR
APOSPC302226-5G APOSPC302226-1G
2'-Chlor-3'-(trifluormethyl)acetophenon 97%
2'-Chlor-3'-(trifluormethyl)acetophenon
Formel: C₉H₆ClF₃O
Siedepunkt: 90…91 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09832253
CAS-Nummer: 129322-82-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR