2-Chloro-6-(trifluoromethoxy)aniline 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-6-(trifluoromethoxy)aniline
PC302230-1G
PC302230-5G
PC302230-10G
PC302230-25G
PC302230-100G
APOSPC302230-1GEA
92.8
EUR
APOSPC302230-1G
APOSPC302230-5G
APOSPC302230-10G
APOSPC302230-25G
APOSPC302230-100G
2-Chloro-6-(trifluoromethoxy)aniline 98%
2-Chloro-6-(trifluoromethoxy)aniline
Formel:
C₇H₅ClF₃NO Molecular Weight: 211,57 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07779302 CAS-Nummer: 1261673-70-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...