2-Chlor-6-(trifluormethyl)benzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Chlor-α,α,α-trifluor-o-toluylsäure
PC302231-5G
PC302231-1G
PC302231-10G
APOSPC302231-5GEA
63.8
EUR
APOSPC302231-5G
APOSPC302231-1G
APOSPC302231-10G
2-Chlor-6-(trifluormethyl)benzoesäure 98%
2-Chloro-6-(trifluoromethyl)benzoic acid
Formel:
C₈H₄ClF₃O₂ Molecular Weight: 224,57 g/mol Schmelzpunkt: 124…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06208254 CAS-Nummer: 2376-00-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...