2-Methoxy-4-(trifluormethyl)benzylamin 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC302251-1G
PC302251-5G
PC302251-10G
APOSPC302251-1GEA
159
EUR
APOSPC302251-1G
APOSPC302251-5G
APOSPC302251-10G
2-Methoxy-4-(trifluormethyl)benzylamin 97%
2-Methoxy-4-(trifluormethyl)benzylamin
Formel:
C₉H₁₀F₃NO Molecular Weight: 205,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06213016 CAS-Nummer: 771581-99-8 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...