3-Chlor-4-(trifluormethoxy)benzylamin 98%

Lieferant: Apollo Scientific
Gefahr

PC302286-1G PC302286-5G
APOSPC302286-1GEA 116 EUR
APOSPC302286-1G APOSPC302286-5G
3-Chlor-4-(trifluormethoxy)benzylamin 98%
3-Chlor-4-(trifluormethoxy)benzylamin
Formel: C₈H₇ClF₃NO
Molecular Weight: 225.60 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06212980
CAS-Nummer: 771581-60-3
UN: 2735
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR