3-Chlor-5-fluor-4-methoxyphenol 97%

Lieferant: Apollo Scientific

PC302293-1G PC302293-5G PC302293-10G
APOSPC302293-1GEA 136 EUR
APOSPC302293-1G APOSPC302293-5G APOSPC302293-10G
3-Chlor-5-fluor-4-methoxyphenol 97%
3-Chlor-5-fluor-4-methoxyphenol
Formel: C₇H₆ClFO₂
Molecular Weight: 176,57 g/mol
Schmelzpunkt: 62…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09832264
CAS-Nummer: 1017777-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR