3-Ethoxy-2,6-difluorophenylacetonitrile 97%

Lieferant: Apollo Scientific

PC302297-1G PC302297-5G
APOSPC302297-1GEA 92.8 EUR
APOSPC302297-1G APOSPC302297-5G
3-Ethoxy-2,6-difluorophenylacetonitrile 97%
3-Ethoxy-2,6-difluorophenylacetonitrile
Formel: C₁₀H₉F₂NO
Molecular Weight: 197,18 g/mol
Schmelzpunkt: 50…53 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11519375
CAS-Nummer: 1092461-33-0
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR