4-(Pentafluorothio)phenylacetonitrile

Lieferant: Apollo Scientific

PC302317-1G PC302317-5G
APOSPC302317-1GEA 516 EUR
APOSPC302317-1G APOSPC302317-5G
4-(Pentafluorothio)phenylacetonitrile
4-(Pentafluorothio)phenylacetonitrile
Formel: C₈H₆F₅NS
Molecular Weight: 243,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16652419
CAS-Nummer: 1240257-04-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR