4-n-Butoxy-2,6-difluorobenzyl bromide 97%
Lieferant: Apollo Scientific
Synonyme:
4-Butoxy-2,6-difluorobenzyl bromide
PC302318-1G
PC302318-5G
PC302318-10G
APOSPC302318-1GEA
87
EUR
APOSPC302318-1G
APOSPC302318-5G
APOSPC302318-10G
4-n-Butoxy-2,6-difluorobenzyl bromide 97%
4-n-Butoxy-2,6-difluorobenzyl bromide
Formel:
C₁₁H₁₃BrF₂O Molecular Weight: 279,12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22201021 CAS-Nummer: 1373921-06-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...