4-Chloro-2-fluoro-3-methoxybenzoyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-chloro-2-fluoro-3-methoxybenzoyl chloride

PC302323-1G PC302323-5G
APOSPC302323-1GEA 92.8 EUR
APOSPC302323-1G APOSPC302323-5G
4-Chloro-2-fluoro-3-methoxybenzoyl chloride 97%
4-Chloro-2-fluoro-3-methoxybenzoyl chloride
Formel: C₈H₅Cl₂FO₂
Molecular Weight: 223,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19687205
CAS-Nummer: 1323966-24-0
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR