4-Chloro-3-ethoxy-2-fluorobenzoyl chloride 97%

Lieferant: Apollo Scientific

Synonyme: 4-chloro-3-ethoxy-2-fluorobenzoyl chloride

PC302327-1G PC302327-5G
APOSPC302327-1GEA 132 EUR
APOSPC302327-1G APOSPC302327-5G
4-Chloro-3-ethoxy-2-fluorobenzoyl chloride 97%
4-Chloro-3-ethoxy-2-fluorobenzoyl chloride
Formel: C₉H₇Cl₂FO₂
Molecular Weight: 237,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19687216
CAS-Nummer: 1323966-42-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR