5-Chlor-2-(trifluormethoxy)benzylalkohol

Lieferant: Apollo Scientific
Achtung

PC302354-1G PC302354-5G
APOSPC302354-1GEA 157 EUR
APOSPC302354-1G APOSPC302354-5G
5-Chlor-2-(trifluormethoxy)benzylalkohol
5-Chlor-2-(trifluormethoxy)benzylalkohol
Formel: C₈H₆ClF₃O₂
Molecular Weight: 226.58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11519357
CAS-Nummer: 874821-52-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR