5-Fluor-2-(trifluormethoxy)anilin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Fluoro-2-(trifluoromethoxy)benzylamine
PC302356-1G
PC302356-5G
PC302356-10G
PC302356-25G
APOSPC302356-1GEA
21.9
EUR
APOSPC302356-1G
APOSPC302356-5G
APOSPC302356-10G
APOSPC302356-25G
5-Fluor-2-(trifluormethoxy)anilin 98%
5-Fluor-2-(trifluormethoxy)anilin
Formel:
C₇H₅F₄NO Molecular Weight: 195,11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16652514 CAS-Nummer: 123572-63-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...