(2,4-Difluor-3-methoxyphenyl)essigsäure 98%
Lieferant: Apollo Scientific
PC302376-1G
PC302376-5G
PC302376-10G
APOSPC302376-1GEA
148
EUR
APOSPC302376-1G
APOSPC302376-5G
APOSPC302376-10G
(2,4-Difluor-3-methoxyphenyl)essigsäure 98%
(2,4-Difluor-3-methoxyphenyl)essigsäure
Formel:
(C₆H₂F₂(OCH₃))CH₂COOH Molecular Weight: 186.16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115939 CAS-Nummer: 886499-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...