2,6-Dichlor-3-(trifluormethyl)benzoesäure 97%

Lieferant: Apollo Scientific

PC302379-5G PC302379-10G PC302379-1G
APOSPC302379-5GEA 203 EUR
APOSPC302379-5G APOSPC302379-10G APOSPC302379-1G
2,6-Dichlor-3-(trifluormethyl)benzoesäure 97%
2,6-Dichloro-3-(trifluoromethyl)benzoic acid
Formel: C₈H₃Cl₂F₃O₂
Molecular Weight: 259.01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09743917
CAS-Nummer: 25922-41-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR