2-Brom-4-chlor-6-fluoranisol 98%

Lieferant: Apollo Scientific
Achtung

PC302381-5G PC302381-1G
APOSPC302381-5GEA 161 EUR
APOSPC302381-5G APOSPC302381-1G
2-Brom-4-chlor-6-fluoranisol 98%
2-Brom-4-chlor-6-fluoranisol
Formel: C₇H₅BrClFO
Molecular Weight: 239,47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115953
CAS-Nummer: 886499-78-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR