2-Brom-6-chlor-4-fluoranisol 98%

Lieferant: Apollo Scientific
Achtung

PC302382-5G PC302382-10G PC302382-1G
APOSPC302382-5GEA 130 EUR
APOSPC302382-5G APOSPC302382-10G APOSPC302382-1G
2-Brom-6-chlor-4-fluoranisol 98%
2-Brom-6-chlor-4-fluoranisol
Formel: C₇H₅BrClFO
Molecular Weight: 239.47 g/mol
Schmelzpunkt: 40…41 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115952
CAS-Nummer: 222712-93-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR