(3,5-Difluor-2-methoxyphenyl)essigsäure 98%
Lieferant: Apollo Scientific
PC302393-1G
PC302393-5G
APOSPC302393-1GEA
105
EUR
APOSPC302393-1G
APOSPC302393-5G
(3,5-Difluor-2-methoxyphenyl)essigsäure 98%
(3,5-Difluor-2-methoxyphenyl)essigsäure
Formel:
(C₆H₂F₂(OCH₃))CH₂COOH Molecular Weight: 186,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660333 CAS-Nummer: 886761-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...