3-Chlor-4-(trifluormethyl)benzaldehyd 98%

Lieferant: Apollo Scientific
Achtung

PC302394-5G PC302394-10G PC302394-1G
APOSPC302394-5GEA 65.4 EUR
APOSPC302394-5G APOSPC302394-10G APOSPC302394-1G
3-Chlor-4-(trifluormethyl)benzaldehyd 98%
3-Chloro-4-(trifluoromethyl)benzaldehyde
Formel: C₈H₄ClF₃O
Molecular Weight: 208,57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095197
CAS-Nummer: 83279-38-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR